2,4,5-Trifluorphenylisothiocyanat
Lieferant: FLUOROCHEM
009601-1G
FLUO009601-1GEA
203
EUR
FLUO009601-1G
2,4,5-Trifluorphenylisothiocyanat
2,4,5-Trifluorphenylisothiocyanat
Formel:
C₇H₂F₃NS Molecular Weight: 189,16 g/mol |
CAS-Nummer:
932710-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...