2,3,6-Trifluorphenylisothiocyanat

Lieferant: FLUOROCHEM

009602-1G
FLUO009602-1GEA 335 EUR
FLUO009602-1G
2,3,6-Trifluorphenylisothiocyanat
2,3,6-Trifluorphenylisothiocyanat
Formel: C₇H₂F₃NS
CAS-Nummer: 362690-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR