2,3,4-Trifluorphenylisothiocyanat
Lieferant: FLUOROCHEM
009604-1G
FLUO009604-1GEA
97.2
EUR
FLUO009604-1G
2,3,4-Trifluorphenylisothiocyanat
2,3,4-Trifluorphenylisothiocyanat
Formel:
C₇H₂F₃NS Molecular Weight: 189,16 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
119474-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...