2,3-Difluorphenylisothiocyanat

Lieferant: FLUOROCHEM

009611-1G
FLUO009611-1GEA 317 EUR
FLUO009611-1G
2,3-Difluorphenylisothiocyanat
2,3-Difluorphenylisothiocyanat
Formel: C₇H₃F₂NS
CAS-Nummer: 363179-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR