4-Methoxy-N-methylanilin

Lieferant: FLUOROCHEM

Synonyme: N-Methyl-p-anisidine

009653-10G
FLUO009653-10GEA 254 EUR
FLUO009653-10G
4-Methoxy-N-methylanilin
4-Methoxy-N-methylanilin
Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
MDL: MFCD00008399
CAS-Nummer: 5961-59-1
EINECS: 227-735-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR