Mono(trimethylsilyl)phosphit

Lieferant: FLUOROCHEM

009689-10G
FLUO009689-10GEA 0 EUR
FLUO009689-10G
Mono(trimethylsilyl)phosphit
Mono(trimethylsilyl)phosphit
Formel: C₃H₁₁O₃PSi
MDL: MFCD00800498
CAS-Nummer: 91076-68-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR