4-Chlorphenacylthiocyanat
Lieferant: FLUOROCHEM
Synonyme:
1-(4-Chlorophenyl)-2-thiocyanatoethanone, 2-(4-Chlorophenyl)-2-oxoethyl thiocyanate
009708-10G
FLUO009708-10GEA
0
EUR
FLUO009708-10G
4-Chlorphenacylthiocyanat
4-Chlorphenacylthiocyanat
Formel:
C₉H₆ClNOS Molecular Weight: 211,668 g/mol |
MDL:
MFCD00019771 CAS-Nummer: 19339-59-4 EINECS: 242-972-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...