5-Amino-3-phenyl-1,2,4-thiadiazol

Lieferant: FLUOROCHEM

Synonyme: 3-Phenyl-1,2,4-thiadiazol-5-amine

009735-10G
FLUO009735-10GEA 0 EUR
FLUO009735-10G
5-Amino-3-phenyl-1,2,4-thiadiazol
5-Amino-3-phenyl-1,2,4-thiadiazole
Formel: C₈H₇N₃S
Molecular Weight: 177,23 g/mol
MDL: MFCD00003159
CAS-Nummer: 17467-15-1
EINECS: 241-485-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR