4-Fluor-2-nitrobenzoesäure

Lieferant: FLUOROCHEM

CFF1135. 009748-1G
CHMPCFF1135.EA 0 EUR
CHMPCFF1135. FLUO009748-1G
4-Fluor-2-nitrobenzoesäure
4-Fluoro-2-nitrobenzoic acid
Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
MDL: MFCD00024300
CAS-Nummer: 394-01-4
EINECS: 206-890-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR