1-(3,4-Dichlorphenyl)piperazin
Lieferant: FLUOROCHEM
|
Achtung
|
009768-1G
009768-5G
009768-25G
FLUO009768-1GEA
0
EUR
FLUO009768-1G
FLUO009768-5G
FLUO009768-25G
1-(3,4-Dichlorphenyl)piperazin
1-(3,4-Dichlorphenyl)piperazin
Formel:
C₁₀H₁₂Cl₂N₂ Molecular Weight: 231.12 g/mol Siedepunkt: 370 Schmelzpunkt: 62…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023128 CAS-Nummer: 57260-67-0 EINECS: 260-652-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...