1-(2-Hydroxyphenyl)piperazin
Lieferant: FLUOROCHEM
Synonyme:
o-(1-Piperazinyl)phenol
009795.10G
009795-50G
FLUO009795.10GEA
68.1
EUR
FLUO009795.10G
FLUO009795-50G
1-(2-Hydroxyphenyl)piperazin
1-(2-Hydroxyphenyl)piperazin
Formel:
C₁₀H₁₄N₂O Siedepunkt: 318 °C (1013 hPa) Schmelzpunkt: 127 °C |
MDL:
MFCD00190246 CAS-Nummer: 1011-17-2 EINECS: 213-782-5 UN: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...