Ethyl-3-piperidincarboxylat

Lieferant: FLUOROCHEM

009838-5G 009838-25G 009838-100G
FLUO009838-5GEA 0 EUR
FLUO009838-5G FLUO009838-25G FLUO009838-100G
Ethyl-3-piperidincarboxylat
Ethyl-3-piperidincarboxylat
Formel: C₈H₁₅NO₂
MDL: MFCD00005991
CAS-Nummer: 5006-62-2
EINECS: 225-681-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR