2-Methyl-3-(trifluormethyl)benzoesäure

Lieferant: FLUOROCHEM

009858-1G 009858-5G
FLUO009858-1GEA 66.5 EUR
FLUO009858-1G FLUO009858-5G
2-Methyl-3-(trifluormethyl)benzoesäure
2-Methyl-3-(trifluoromethyl)benzoic acid
Formel: C₉H₇F₃O₂
MDL: MFCD01631588
CAS-Nummer: 62089-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR