5-Nitro-2-benzofurancarbonsäure
Lieferant: FLUOROCHEM
009951-1G
FLUO009951-1GEA
76.2
EUR
FLUO009951-1G
5-Nitro-2-benzofurancarbonsäure
5-Nitrobenzofuran-2-carboxylic acid
Formel:
C₉H₅NO₅ Molecular Weight: 207,1397 g/mol |
MDL:
MFCD00060513 CAS-Nummer: 10242-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...