1-(2-(Ammoniooxy)ethoxy)-3-(trifluormethyl)benzolchlorid
Lieferant: FLUOROCHEM
Synonyme:
O-{2-[3-(Trifluoromethyl)phenoxy]ethyl}hydroxylamine hydrochloride
010008-1G
FLUO010008-1GEA
519
EUR
FLUO010008-1G
1-(2-(Ammoniooxy)ethoxy)-3-(trifluormethyl)benzolchlorid
1-(2-(Ammoniooxy)ethoxy)-3-(trifluormethyl)benzolchlorid
Formel:
C₉H₁₁ClF₃NO₂ |
MDL:
MFCD01114594 CAS-Nummer: 113211-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...