1-[(Ammoniooxy)methyl]-3-(trifluormethyl)benzolchlorid
Lieferant: FLUOROCHEM
Synonyme:
O-[3-(Trifluoromethyl)benzyl]hydroxylamine hydrochloride
010011-1G
FLUO010011-1GEA
0
EUR
FLUO010011-1G
1-[(Ammoniooxy)methyl]-3-(trifluormethyl)benzolchlorid
1-[(Ammoniooxy)methyl]-3-(trifluormethyl)benzolchlorid
Formel:
C₈H₉ClF₃NO |
MDL:
MFCD01114571 CAS-Nummer: 15256-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...