2-(4-Fluorphenoxy)benzaldehyd

Lieferant: FLUOROCHEM

010111-1G
FLUO010111-1GEA 201 EUR
FLUO010111-1G
2-(4-Fluorphenoxy)benzaldehyd
2-(4-Fluorophenoxy)benzaldehyde
Formel: C₁₃H₉FO₂
Molecular Weight: 216,21 g/mol
MDL: MFCD01568770
CAS-Nummer: 320423-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR