4-(3-Fluorpropoxy)benzaldehyd

Lieferant: FLUOROCHEM

010124-1G
FLUO010124-1GEA 0 EUR
FLUO010124-1G
4-(3-Fluorpropoxy)benzaldehyd
4-(3-Fluoropropoxy)benzaldehyde
Formel: C₁₀H₁₁FO₂
MDL: MFCD02090999
CAS-Nummer: 400825-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR