1,1,1-Trifluor-3-(phenethylamino)-2-propanol
Lieferant: FLUOROCHEM
Synonyme:
3-Phenethylamino-1,1,1-trifluoropropan-2-ol
010178-500MG
FLUO010178-500MGEA
0
EUR
FLUO010178-500MG
1,1,1-Trifluor-3-(phenethylamino)-2-propanol
1,1,1-Trifluor-3-(phenethylamino)-2-propanol
Formel:
C₁₁H₁₄F₃NO |
MDL:
MFCD03001278 CAS-Nummer: 400878-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...