6-Fluorgramin

Lieferant: FLUOROCHEM

Synonyme: 1-(6-Fluoro-1H-indol-3-yl)-N,N-dimethylmethanamine

010187-1G
FLUO010187-1GEA 63.9 EUR
FLUO010187-1G
6-Fluorgramin
6-Fluorgramin
Formel: C₁₁H₁₃FN₂
Molecular Weight: 192,24 g/mol
MDL: MFCD00056918
CAS-Nummer: 343-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR