6-Fluor-3-indolylessigsäure
Lieferant: FLUOROCHEM
Synonyme:
6-Fluorindol-3-ylessigsäure, 6-Fluorindol-3-essigsäure
010189-1G
FLUO010189-1GEA
293
EUR
FLUO010189-1G
6-Fluor-3-indolylessigsäure
6-Fluor-3-indolylessigsäure
Formel:
C₁₀H₈FNO₂ |
MDL:
MFCD01074503 CAS-Nummer: 443-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...