5-Fluortryptamin Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: 2-(5-Fluoro-1H-indol-3-yl)ethanamine hydrochloride

010196-1G
FLUO010196-1GEA 0 EUR
FLUO010196-1G
5-Fluortryptamin Hydrochlorid
5-Fluortryptamin Hydrochlorid
Formel: C₁₀H₁₂ClFN₂
MDL: MFCD00051081
CAS-Nummer: 2711-58-2
EINECS: 220-307-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR