6-Fluortryptamin

Lieferant: FLUOROCHEM

010197-1G
FLUO010197-1GEA 0 EUR
FLUO010197-1G
6-Fluortryptamin
6-Fluortryptamin
Formel: C₁₀H₁₁FN₂
Molecular Weight: 178,21 g/mol
MDL: MFCD00044761
CAS-Nummer: 575-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR