6-Fluortryptamin Hydrochlorid

Lieferant: FLUOROCHEM

010198-1G
FLUO010198-1GEA 0 EUR
FLUO010198-1G
6-Fluortryptamin Hydrochlorid
6-Fluortryptamin Hydrochlorid
Formel: C₁₀H₁₂ClFN₂
Molecular Weight: 214,67 g/mol
MDL: MFCD00012687
CAS-Nummer: 55206-24-1
EINECS: 259-533-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR