4-Fluor-DL-tryptophan

Lieferant: FLUOROCHEM

Synonyme: 2-Amino-3-(4-fluoro-1H-indol-3-yl)propanoic acid, H-DL-Trp(4-F)-OH

010200-1G
FLUO010200-1GEA 1050 EUR
FLUO010200-1G
4-Fluor-DL-tryptophan
4-Fluor-DL-tryptophan
Formel: C₁₁H₁₁FN₂O₂
Molecular Weight: 222,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039689
CAS-Nummer: 25631-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR