6-Fluor-DL-tryptophan

Lieferant: FLUOROCHEM

Synonyme: 2-Amino-3-(6-fluoro-1H-indol-3-yl)propanoic acid, 6-Fluoro-DL-tryptophan

010201-1G
FLUO010201-1GEA 342 EUR
FLUO010201-1G
6-Fluor-DL-tryptophan
6-Fluor-DL-tryptophan
Formel: C₁₁H₁₁FN₂O₂
Molecular Weight: 222,22 g/mol
Schmelzpunkt: 280 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00005653
CAS-Nummer: 7730-20-3
EINECS: 231-788-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR