2,6-Dinitro-4-(trifluormethyl)anilin
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
4-Amino-3,5-dinitro-α,α,α-trifluortoluol, 2,6-Dinitro-α,α,α-trifluor-p-toluidin
010203-1G
FLUO010203-1GEA
87.9
EUR
FLUO010203-1G
2,6-Dinitro-4-(trifluormethyl)anilin
2,6-Dinitro-4-(trifluormethyl)anilin
Formel:
C₇H₄F₃N₃O₄ Molecular Weight: 251,12 g/mol |
MDL:
MFCD00024260 CAS-Nummer: 445-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...