4-Brom-3,5-difluorbenzotrifluorid
Lieferant: FLUOROCHEM
Synonyme:
4-Brom-3,5-difluor-α,α,α-trifluortoluol, 1-Brom-2,6-difluor-4-(trifluormethyl)benzol
010204-1G
FLUO010204-1GEA
0
EUR
FLUO010204-1G
4-Brom-3,5-difluorbenzotrifluorid
4-Brom-3,5-difluorbenzotrifluorid
Formel:
C₇H₂BrF₅ Molecular Weight: 260,99 g/mol |
MDL:
MFCD03093034 CAS-Nummer: 499238-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...