3'-Brom-4'-fluoracetophenon

Lieferant: FLUOROCHEM

010205-10G 010205-100G 010205-25G
FLUO010205-10GEA 43.1 EUR
FLUO010205-10G FLUO010205-100G FLUO010205-25G
3'-Brom-4'-fluoracetophenon
3'-Brom-4'-fluoracetophenon
Formel: C₈H₆BrFO
Molecular Weight: 217.04 g/mol
Siedepunkt: 240
Schmelzpunkt: 57
MDL: MFCD00042466
CAS-Nummer: 1007-15-4
EINECS: 213-750-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR