2-Chlor-4-nitrobenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 2-Chlor-4-nitrobenzotrifluorid, 1-Chlor-3-nitro-6-(trifluormethyl)benzol

010206-1G
FLUO010206-1GEA 0 EUR
FLUO010206-1G
2-Chlor-4-nitrobenzotrifluorid
2-Chlor-4-nitrobenzotrifluorid
Formel: C₇H₃ClF₃NO₂
Molecular Weight: 225,55 g/mol
MDL: MFCD00171532
CAS-Nummer: 151504-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR