7-Fluorisatin

Lieferant: FLUOROCHEM
Achtung

CFF1118. 010208-1G 010208-25G 010208-5G
CHMPCFF1118.EA 0 EUR
CHMPCFF1118. FLUO010208-1G FLUO010208-25G FLUO010208-5G
7-Fluorisatin
7-Fluorisatin
Formel: C₈H₄FNO₂
MDL: MFCD01569508
CAS-Nummer: 317-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR