α-(Trifluormethyl)benzhydrol

Lieferant: FLUOROCHEM

Synonyme: 2,2,2-Trifluoro-1,1-diphenylethanol

010212-1G
FLUO010212-1GEA 0 EUR
FLUO010212-1G
α-(Trifluormethyl)benzhydrol
α-(Trifluormethyl)benzhydrol
Formel: C₁₄H₁₁F₃O
Molecular Weight: 252,230 g/mol
MDL: MFCD00021800
CAS-Nummer: 379-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR