2,5-Bis(trifluormethyl)benzoesäure

Lieferant: FLUOROCHEM

010242-1G 010242-5G
FLUO010242-1GEA 66.8 EUR
FLUO010242-1G FLUO010242-5G
2,5-Bis(trifluormethyl)benzoesäure
2,5-Bis(trifluormethyl)benzoesäure
Formel: C₉H₄F₆O₂
MDL: MFCD00013249
CAS-Nummer: 42580-42-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR