1-Brom-2,3,3,3-tetrafluorpropen
Lieferant: FLUOROCHEM
010260-1G
FLUO010260-1GEA
0
EUR
FLUO010260-1G
1-Brom-2,3,3,3-tetrafluorpropen
1-Bromo-2,3,3,3-tetrafluoropropene
Formel:
C₃HBrF₄ |
MDL:
MFCD03095423 CAS-Nummer: 666-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...