1,2-Dibromperfluorheptan

Lieferant: FLUOROCHEM

010289-5G 010289-25G
FLUO010289-5GEA 0 EUR
FLUO010289-5G FLUO010289-25G
1,2-Dibromperfluorheptan
1,2-Dibromperfluorheptan
Formel: C₇Br₂F₁₄
MDL: MFCD03094409
CAS-Nummer: 678-32-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR