1,8-Dichlorhexadecafluoroctan

Lieferant: FLUOROCHEM

010296-25G 010296-100G 010296-5G
FLUO010296-25GEA 0 EUR
FLUO010296-25G FLUO010296-100G FLUO010296-5G
1,8-Dichlorhexadecafluoroctan
1,8-Dichlorohexadecafluorooctane
Formel: C₈Cl₂F₁₆
MDL: MFCD00155747
CAS-Nummer: 647-25-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR