1,4-Difluorbutan
Lieferant: FLUOROCHEM
010297-1G
010297-5G
FLUO010297-1GEA
320
EUR
FLUO010297-1G
FLUO010297-5G
1,4-Difluorbutan
1,4-Difluorobutane
Formel:
C₄H₈F₂ |
MDL:
MFCD01740905 CAS-Nummer: 372-90-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...