1,1,2,3,3,3-Hexafluorpropyldifluormethylether

Lieferant: FLUOROCHEM

010320-1G 010320-5G
FLUO010320-1GEA 57.9 EUR
FLUO010320-1G FLUO010320-5G
1,1,2,3,3,3-Hexafluorpropyldifluormethylether
1,1,2,3,3,3-Hexafluorpropyldifluormethylether
Formel: C₄H₂F₈O
MDL: MFCD03094220
CAS-Nummer: 56860-85-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR