2-Iod-3,3,3-trifluorpropen
Lieferant: FLUOROCHEM
010325-1G
010325-5G
FLUO010325-1GEA
0
EUR
FLUO010325-1G
FLUO010325-5G
2-Iod-3,3,3-trifluorpropen
2-Iodo-3,3,3-trifluoropropene
Formel:
C₃H₂F₃I Molecular Weight: 221.95 g/mol |
MDL:
MFCD02183571 CAS-Nummer: 107070-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...