(2-Nitro-4-(trifluormethyl)phenyl)essigsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2-[2-Nitro-4-(trifluoromethyl)phenyl]acetic acid
010345-1G
010345-5G
FLUO010345-1GEA
36.9
EUR
FLUO010345-1G
FLUO010345-5G
(2-Nitro-4-(trifluormethyl)phenyl)essigsäure
(2-Nitro-4-(trifluormethyl)phenyl)essigsäure
Formel:
C₉H₆F₃NO₄ Molecular Weight: 249.15 g/mol |
MDL:
MFCD00091534 CAS-Nummer: 1735-91-7 EINECS: 217-081-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...