2-Chlorisonicotinsäure
Lieferant: FLUOROCHEM
Synonyme:
2-Chlorpyridin-4-carbonsäure, 2-Chlor-4-pyridincarbonsäure
CFF134.2.5
CFF134.5
010413-5G
010413-25G
010413-100G
CHMPCFF134.2.5EA
348
EUR
CHMPCFF134.2.5
CHMPCFF134.5
FLUO010413-5G
FLUO010413-25G
FLUO010413-100G
2-Chlorisonicotinsäure
2-Chlorisonicotinsäure
Formel:
C₆H₄ClNO₂ |
MDL:
MFCD00191402 CAS-Nummer: 6313-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...