4,4,5,5,6,6,6-Heptafluor-1-hexanol

Lieferant: FLUOROCHEM

Synonyme: 4,4,5,5,6,6,6-Heptafluorohexan-1-ol

010475-25G 010475-5G
FLUO010475-25GEA 0 EUR
FLUO010475-25G FLUO010475-5G
4,4,5,5,6,6,6-Heptafluor-1-hexanol
4,4,5,5,6,6,6-Heptafluoro-1-hexanol
Formel: C₆H₇F₇O
MDL: MFCD00077554
CAS-Nummer: 679-02-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR