1H,1H,13H-Tetracosafluor-1-tridecanol
Lieferant: FLUOROCHEM
010523-25G
FLUO010523-25GEA
0
EUR
FLUO010523-25G
1H,1H,13H-Tetracosafluor-1-tridecanol
1H,1H,13H-Tetracosafluor-1-tridecanol
Formel:
C₁₃H₄OF₂₄ |
MDL:
MFCD00155920 CAS-Nummer: 423-72-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...