3-(1,1,2,2-Tetrafluorethoxy)phenol

Lieferant: FLUOROCHEM

010534-25G
FLUO010534-25GEA 0 EUR
FLUO010534-25G
3-(1,1,2,2-Tetrafluorethoxy)phenol
3-(1,1,2,2-Tetrafluorethoxy)phenol
Formel: C₈H₆F₄O₂
MDL: MFCD00153276
CAS-Nummer: 53997-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR