1,1,2-Trifluordecen

Lieferant: FLUOROCHEM

010553-25G
FLUO010553-25GEA 0 EUR
FLUO010553-25G
1,1,2-Trifluordecen
1,1,2-Trifluordecen
Formel: C₁₀H₁₇F₃
MDL: MFCD00156047
CAS-Nummer: 692-05-7
EINECS: 206-590-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR