2-Aminoethyldihydrogenphosphat
Lieferant: FLUOROCHEM
Synonyme:
O-Phosphorylethanolamine
010650-25G
010650-100G
FLUO010650-25GEA
65
EUR
FLUO010650-25G
FLUO010650-100G
2-Aminoethyldihydrogenphosphat
2-Aminoethyldihydrogenphosphat
Formel:
C₂H₈NO₄P Molecular Weight: 141.0630 g/mol |
MDL:
MFCD00008178 CAS-Nummer: 1071-23-4 EINECS: 213-988-5 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...