6-Amino-2-naphthoesäure

Lieferant: FLUOROCHEM

Synonyme: 6-Amino-2-naphthoic acid, 6-Amino-2-naphthalenecarboxylic acid, 6-Amino-2-naphthoic Acid

010652-5G 010652-25G 010652-1G
FLUO010652-5GEA 483 EUR
FLUO010652-5G FLUO010652-25G FLUO010652-1G
6-Amino-2-naphthoesäure
6-Amino-2-naphthoesäure
Formel: C₁₁H₉NO₂
MDL: MFCD01861831
CAS-Nummer: 116668-47-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR