3-Amino-2-naphthoesäure
Lieferant: FLUOROCHEM
Synonyme:
3-Amino-2-naphthalincarbonsäure, 3-Aminonaphthalin-2-carbonsäure
010658-25G
FLUO010658-25GEA
476
EUR
FLUO010658-25G
3-Amino-2-naphthoesäure
3-Amino-2-naphthoesäure
Formel:
C₁₁H₉NO₂ Siedepunkt: 383 °C (1013 hPa) Schmelzpunkt: 216…217 °C |
MDL:
MFCD00004115 CAS-Nummer: 5959-52-4 EINECS: 227-726-2 Merck Index: 13,00451 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...