3-Fluor-p-anisidin

Lieferant: FLUOROCHEM

010679-1G 010679-5G 010679 010679-25G
FLUO010679-1GEA 31.1 EUR
FLUO010679-1G FLUO010679-5G FLUO010679 FLUO010679-25G
3-Fluor-p-anisidin
3-Fluor-p-anisidin
Formel: C₇H₈FNO
MDL: MFCD00075040
CAS-Nummer: 366-99-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR