2-Fluor-4-iodanilin
Lieferant: FLUOROCHEM
010682-25G
010682-100G
FLUO010682-25GEA
45.8
EUR
FLUO010682-25G
FLUO010682-100G
2-Fluor-4-iodanilin
2-Fluor-4-iodanilin
Formel:
C₆H₅FIN Siedepunkt: 259 °C (1013 hPa) Schmelzpunkt: 52…54 °C |
MDL:
MFCD00011738 CAS-Nummer: 29632-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...